Assay Details
| Gene Symbol: LOC100996573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003846334.4 | XP_003846382.2 | 1 | 3761 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003846334.4 transcripts.
Set Membership: |
|

LOC100996573
Genomic Map