Assay Details
| Gene Symbol: ZC3H10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303124.1 | NP_001290053.1 | 3 | 1074 | 129 | |
| NM_001303125.1 | NP_001290054.1 | 3 | 1043 | 129 | ||
| NM_032786.2 | NP_116175.1 | 3 | 934 | 129 | ||
| GenBank mRNA | AK027357.1 | - | 3 | 908 | 129 | |
| BC018708.1 | - | 3 | 896 | 4111500 | 129 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000257940.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000546903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000549973.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000551880.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | ENST00000552345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303124.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_001303125.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012645.hg.1 | NM_032786.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000549438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010893.hg.1 | ENST00000550947.1 | Ensembl |

ZC3H10
Genomic Map