Assay Details
| Gene Symbol: CAPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005269192.1 | XP_005269249.1 | 1 - 2 | 62 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005269192.1 transcripts.
Set Membership: |
|

CAPS2
Genomic Map