Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000540490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000544251.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
ENST00000546180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012391.hg.1 |
NR_120451.1 |
Refseq |