Assay Details
| Gene Symbol: UHRF1BP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017019047.1 | XP_016874536.1 | 21 | 6656 | 85 | |
| GenBank mRNA | BC026304.1 | - | 1 | 1122 | 85 | |
More Information
Set Membership: |
|

UHRF1BP1L
Genomic Map