Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000361483.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000547838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000548163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
ENST00000549321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177996.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_001177997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011931.hg.1 |
NM_144671.4 |
Refseq |