Assay Details
| Gene Symbol: LOC102724050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120488.1 | 2 - 3 | 244 | 97 | ||
| GenBank mRNA | AI809671.1 | - | 1 - 2 | 97 | ||
| DA757660.1 | - | 2 - 3 | 125 | 97 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000548617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000550474.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | ENST00000552785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120487.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007763.hg.1 | NR_120488.1 | Refseq |

LOC102724050
Genomic Map