Assay Details
| Gene Symbol: PTPRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017019273.1 | XP_016874762.1 | 1 - 2 | 324 | 82 | |
| XM_017019274.1 | XP_016874763.1 | 1 - 2 | 325 | 82 | ||
| XM_017019275.1 | XP_016874764.1 | 1 - 2 | 325 | 82 | ||
More Information
Set Membership: |
|

PTPRQ
Genomic Map