Assay Details
| Gene Symbol: P3H3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014262.4 | NP_055077.2 | 15 | 2442 | 109 | |
| GenBank mRNA | AB209320.1 | - | 13 | 2209 | 109 | |
| AJ430349.1 | - | 16 | 2671 | 109 | ||
| AK025976.1 | - | 15 | 2409 | 109 | ||
| AK096771.1 | - | 1 | 1966 | 109 | ||
| AK126766.1 | - | 16 | 3004 | 109 | ||
| BC017217.2 | - | 15 | 1983 | 109 | ||
| BC067251.1 | - | 15 | 2140 | 109 | ||
| BC080630.1 | - | 15 | 2215 | 109 | ||
| BQ009002.1 | - | 1 | 109 | |||
| U47926.1 | - | 14 | 1907 | 109 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AX772987.1 | - | 1 | Chr. 12 - 6837955 - 6839847 [-] on Build GRCh38 | 109 | ||
More Information
Set Membership: |
|

P3H3
Genomic Map