Assay Details
| Gene Symbol: SYTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193308.1 | NP_001180237.1 | 2 - 3 | 389 | 83 | |
| NM_032872.2 | NP_116261.1 | 2 - 3 | 389 | 83 | ||
| XM_005246022.3 | XP_005246079.1 | 2 - 3 | 544 | 83 | ||
| XM_006710990.1 | XP_006711053.1 | 2 - 3 | 544 | 83 | ||
| XM_006710991.1 | XP_006711054.1 | 2 - 3 | 340 | 83 | ||
| GenBank mRNA | AK027902.1 | - | 2 - 3 | 359 | 83 | |
| AK074154.1 | - | 1 - 2 | 367 | 83 | ||
| AK096437.1 | - | 2 - 3 | 541 | 83 | ||
| AK293038.1 | - | 1 - 2 | 655 | 83 | ||
| AY037157.1 | - | 2 - 3 | 316 | 83 | ||
| BC015764.1 | - | 1 - 2 | 132 | 4851468 | 83 | |
| BC035725.1 | - | 2 - 3 | 296 | 5722683 | 83 | |
| DA435960.1 | - | 2 - 3 | 389 | 83 | ||
| DA948740.1 | - | 2 - 3 | 366 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC015764.1, XM_006710991.1 transcripts.
Set Membership: |
|

SYTL1
Genomic Map