Assay Details
| Gene Symbol: SIGLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173635.2 | NP_775906.1 | 3 - 4 | 682 | 96 | |
| XM_005258799.4 | XP_005258856.1 | 3 - 4 | 973 | 96 | ||
| XM_011526821.2 | XP_011525123.1 | 3 - 4 | 595 | 96 | ||
| GenBank mRNA | AK097554.1 | - | 3 - 4 | 682 | 96 | |
| BC126357.1 | - | 2 - 3 | 463 | 8992073 | 96 | |
| BC126359.1 | - | 2 - 3 | 463 | 8992075 | 96 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000316401.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000593968.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000596117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597610.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000597824.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000599553.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000601727.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | ENST00000614422.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_001308437.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NM_173635.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008634.hg.1 | NR_102324.1 | Refseq |

SIGLECL1
Genomic Map