Assay Details
| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097590.1 | - | 3 - 4 | 636 | 100 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |

ZNF528-AS1
Genomic Map