Assay Details
| Gene Symbol: FOSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114171.1 | NP_001107643.1 | 1 - 2 | 723 | 118 | |
| NM_006732.2 | NP_006723.2 | 1 - 2 | 723 | 118 | ||
| XM_005258691.1 | XP_005258748.1 | 1 - 2 | 722 | 118 | ||
| GenBank mRNA | AK225070.1 | - | 1 - 2 | 723 | 118 | |
| AK292720.1 | - | 1 - 3 | 322 | 118 | ||
| BC036724.1 | - | 1 - 3 | 702 | 5212854 | 118 | |
| CR542101.1 | - | 1 - 3 | 131 | 118 | ||
| EU178109.1 | - | 1 - 3 | 724 | 118 | ||
| EU178110.1 | - | 1 - 3 | 724 | 118 | ||
| EU178113.1 | - | 1 - 3 | 724 | 118 | ||
| L49169.1 | - | 1 - 3 | 724 | 118 | ||
More Information
Set Membership: |
|

FOSB
Genomic Map