Assay Details
| Gene Symbol: MED16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU392493.1 | - | 2 | 155 | 78 | |
| EU392495.1 | - | 2 | 156 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED16| Gene Symbol: MED16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EU392493.1 | - | 2 | 155 | 78 | |
| EU392495.1 | - | 2 | 156 | 78 | ||
Set Membership: |
|