Assay Details
| Gene Symbol: RAVER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133452.2 | NP_597709.2 | 13 | 3493 | 65 | |
| XM_011527671.1 | XP_011525973.1 | 14 | 3412 | 65 | ||
| GenBank mRNA | AB075858.1 | - | 10 | 4597 | 65 | |
| BC037428.1 | - | 10 | 3006 | 65 | ||
| BC037565.1 | - | 11 | 2830 | 65 | ||
More Information
Set Membership: |
|

RAVER1
Genomic Map