Assay Details
Gene Symbol: ZNF526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314033.1 | NP_001300962.1 | 3 | 999 | 116 | |
NM_133444.2 | NP_597701.1 | 3 | 751 | 116 | ||
GenBank mRNA | AB075831.1 | - | 3 | 682 | 116 | |
AK122649.1 | - | 3 | 675 | 116 | ||
BC013013.2 | - | 3 | 673 | 3531734 | 116 | |
JQ313906.1 | - | 1 | 526 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
