Assay Details
| Gene Symbol: ZMIZ1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015429.1 | 1 - 2 | 56 | 94 | ||
| NR_024429.1 | 1 - 2 | 56 | 94 | |||
| NR_024431.2 | 1 - 2 | 56 | 94 | |||
| GenBank mRNA | AK098249.1 | - | 1 - 2 | 501 | 94 | |
| AK126302.1 | - | 2 - 3 | 425 | 94 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000428862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000440151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | ENST00000456353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_015429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024429.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011152.hg.1 | NR_024431.2 | Refseq |

ZMIZ1-AS1
Genomic Map