Assay Details
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173598.4 | NP_775869.3 | 20 | 16277 | 80 | |
XM_011538224.2 | XP_011536526.1 | 20 | 16606 | 80 | ||
XM_011538225.2 | XP_011536527.1 | 20 | 17071 | 80 | ||
XM_011538228.2 | XP_011536530.1 | 18 | 15436 | 80 | ||
XM_017019210.1 | XP_016874699.1 | 15 | 15062 | 80 | ||
GenBank mRNA | BX648958.1 | - | 1 | 5320 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000339824.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000425217.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000543793.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | ENST00000545002.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200012100.hg.1 | NM_173598.4 | Refseq |
