Assay Details
| Gene Symbol: RAB35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167606.1 | NP_001161078.1 | 5 | 1991 | 103 | |
| NM_006861.6 | NP_006852.1 | 6 | 2116 | 103 | ||
| XM_011537779.1 | XP_011536081.1 | 6 | 2053 | 103 | ||
| XM_017018723.1 | XP_016874212.1 | 5 | 1893 | 103 | ||
| GenBank mRNA | AK001309.2 | - | 1 | 918 | 103 | |
| BC015931.2 | - | 6 | 2031 | 3907209 | 103 | |
| BC017753.1 | - | 1 | 617 | 103 | ||
More Information
Set Membership: |
|

RAB35
Genomic Map