Assay Details
Gene Symbol: SPRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318536.1 | NP_001305465.1 | 1 - 2 | 247 | 61 | |
GenBank mRNA | AK313810.1 | - | 1 - 2 | 247 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK313810.1, NM_001318536.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
