Assay Details
| Gene Symbol: GUCY1B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003923.2 | 14 - 15 | 2188 | 89 | ||
| GenBank mRNA | AF038499.2 | - | 14 - 15 | 2037 | 89 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000389600.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000485590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000493639.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000531898.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000532104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | ENST00000533288.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008971.hg.1 | NR_003923.2 | Refseq |

GUCY1B2
Genomic Map