Assay Details
| Gene Symbol: FGF14-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK093430.1 | - | 1 - 2 | 145 | 71 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000418923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000418923.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000606869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376131.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000376143.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000418923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000418923.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000437115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000468052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000606869.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | ENST00000607251.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_004115.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NM_175929.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009650.hg.1 | NR_036486.1 | Refseq |

FGF14-IT1
Genomic Map