Assay Details
| Gene Symbol: FOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK316360.1 | - | 1 - 2 | 129 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK316360.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000250448.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000545425.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000553751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000554607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | ENST00000557418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008981.hg.1 | NM_004496.3 | Refseq |

FOXA1
Genomic Map