Assay Details
| Gene Symbol: IFI27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288959.1 | NP_001275888.1 | 1 - 2 | 65 | 113 | |
| NM_001288960.1 | NP_001275889.1 | 1 - 2 | 65 | 104 | ||
| GenBank mRNA | AY126457.1 | - | 1 - 2 | 59 | 113 | |
| AY126458.1 | - | 1 - 2 | 59 | 104 | ||
| BM756566.1 | - | 1 - 2 | 62 | 104 | ||
| BU928223.1 | - | 1 - 2 | 66 | 113 | ||
| CB993374.1 | - | 1 - 2 | 103 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY126457.1, AY126458.1, NM_001288959.1, NM_001288960.1 transcripts.
Set Membership: |
|

IFI27
Genomic Map