Assay Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310339.1 | - | 2 - 3 | 161 | 105 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000361487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000402703.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000553267.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000554201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000555639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | ENST00000557107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006933.hg.1 | NM_006194.3 | Refseq |

PAX9
Genomic Map