| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
ENST00000541579.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
ENST00000464767.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
ENST00000541579.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010783.hg.1 |
NR_026779.1 |
Refseq |