Assay Details
| Gene Symbol: LINC01550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097943.1 | - | 2 - 3 | 282 | 101 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000499006.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000502187.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000512901.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000554822.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | ENST00000556462.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010785.hg.1 | NR_015430.1 | Refseq |

LINC01550
Genomic Map