Assay Details
| Gene Symbol: IGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123975.1 | - | 5 - 6 | 1150 | 104 | |
| AK125079.1 | - | 4 - 5 | 1113 | 104 | ||
| AK126352.1 | - | 5 - 6 | 1096 | 104 | ||
| BC066594.1 | - | 5 - 6 | 1059 | 104 | ||
| BX538066.1 | - | 6 - 7 | 1045 | 104 | ||
| BX640822.1 | - | 5 - 6 | 1139 | 104 | ||
| BX640854.1 | - | 5 - 6 | 1127 | 104 | ||
| Gene Symbol: IGHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL389978.1 | - | 4 - 5 | 860 | 104 | |
| BC073765.1 | - | 4 - 5 | 1067 | 104 | ||
| BX538077.1 | - | 5 - 6 | 1088 | 104 | ||
| BX640625.1 | - | 5 - 6 | 1110 | 104 | ||
| BX640710.1 | - | 4 - 5 | 1105 | 104 | ||
More Information
Set Membership: |
|

IGH
Genomic Map