Assay Details
| Gene Symbol: YY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003403.4 | NP_003394.1 | 1 | 930 | 109 | |
| GenBank mRNA | BC020324.1 | - | 1 | 525 | 109 | |
| BC037308.1 | - | 1 | 562 | 5261384 | 109 | |
| BC065366.1 | - | 1 | 495 | 5815774 | 109 | |
| M76541.1 | - | 1 | 524 | 109 | ||
| M77698.1 | - | 1 | 690 | 109 | ||
| Z14077.1 | - | 1 | 705 | 109 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
double-strand break repair via homologous recombination
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
spermatogenesis
response to UV-C
negative regulation of gene expression
cell differentiation
cellular response to UV
four-way junction DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
enhancer sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
transcription regulatory region DNA binding
SMAD binding
negative regulation of transcription from RNA polymerase II promoter
double-strand break repair via homologous recombination
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
spermatogenesis
response to UV-C
negative regulation of gene expression
cell differentiation
cellular response to UV
four-way junction DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
enhancer sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
transcription regulatory region DNA binding
SMAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008205.hg.1 | NR_106822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010923.hg.1 | ENST00000427625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010923.hg.1 | ENST00000427625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010923.hg.1 | ENST00000427625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010923.hg.1 | ENST00000427625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000262238.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000553625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554371.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000554804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | ENST00000623799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NM_003403.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008205.hg.1 | NR_106822.1 | Refseq |

YY1
Genomic Map