Assay Details
| Gene Symbol: NTRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017022242.1 | XP_016877731.1 | 15 - 16 | 2745 | 71 | |
| GenBank mRNA | CD370531.1 | - | 2 - 3 | 71 | ||
More Information
Set Membership: |
|

NTRK3
Genomic Map