Assay Details
| Gene Symbol: ACSBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199377.1 | NP_001186306.1 | 2 - 3 | 442 | 84 | |
| NM_015162.4 | NP_055977.3 | 2 - 3 | 442 | 84 | ||
| XM_011521390.2 | XP_011519692.1 | 2 - 3 | 256 | 84 | ||
| XM_017022025.1 | XP_016877514.1 | 2 - 3 | 276 | 84 | ||
| XM_017022026.1 | XP_016877515.1 | 5 - 6 | 1077 | 84 | ||
| GenBank mRNA | AF179481.1 | - | 2 - 3 | 283 | 84 | |
| AK054882.1 | - | 2 - 3 | 292 | 84 | ||
| AK295269.1 | - | 2 - 3 | 260 | 84 | ||
| AK314508.1 | - | 2 - 3 | 442 | 84 | ||
| BC009289.2 | - | 2 - 3 | 241 | 4298937 | 84 | |
| DC401742.1 | - | 2 - 3 | 442 | 84 | ||
More Information
Set Membership: |
|

ACSBG1
Genomic Map