Assay Details
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC143438.1 | - | 7 - 8 | 3854 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559447.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000559847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | ENST00000560792.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009514.hg.1 | NM_022841.5 | Refseq |
