Assay Details
| Gene Symbol: CEMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293298.1 | NP_001280227.1 | 30 | 5383 | 61 | |
| NM_001293304.1 | NP_001280233.1 | 30 | 5323 | 61 | ||
| NM_018689.2 | NP_061159.1 | 29 | 5224 | 61 | ||
| GenBank mRNA | AB033025.1 | - | 21 | 3892 | 61 | |
| AB103330.1 | - | 29 | 5196 | 61 | ||
| AK026540.1 | - | 14 | 2770 | 61 | ||
| AK027058.1 | - | 1 | 887 | 61 | ||
| AL049389.1 | - | 1 | 422 | 61 | ||
| AL359061.1 | - | 17 | 3267 | 61 | ||
| AY007811.1 | - | 10 | 2355 | 61 | ||
| AY581148.1 | - | 30 | 5355 | 61 | ||
| AY581149.1 | - | 30 | 5295 | 61 | ||
| AY585237.1 | - | 29 | 5196 | 61 | ||
More Information
Set Membership: |
|

CEMIP
Genomic Map