Assay Details
Gene Symbol: LOC149351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC036441.1 | - | 1 - 2 | 343 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000435649.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000443802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000455680.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014891.hg.1 | ENST00000606660.1 | Ensembl |
