Assay Details
| Gene Symbol: LOC102606465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC045807.2 | - | 127 | |||
| BX538249.1 | - | 127 | ||||
More Information
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000416329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000446527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000449473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000451213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000453011.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | ENST00000454721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009231.hg.1 | NR_109849.1 | Refseq |

LOC102606465
Genomic Map