Assay Details
| Gene Symbol: RSAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011510414.2 | XP_011508716.1 | 1 - 2 | 136 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011510414.2 transcripts.
Set Membership: |
|

RSAD2
Genomic Map