Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000328434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
ENST00000586680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010747.hg.1 |
NM_001040431.2 |
Refseq |