Assay Details
| Gene Symbol: ZDHHC8P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003950.1 | 6 | 2037 | 80 | ||
| GenBank mRNA | AK092530.1 | - | 5 | 1866 | 80 | |
| AK092581.1 | - | 3 | 1626 | 80 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000424928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000424928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000441664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000441664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000255890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000418264.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000420968.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000433168.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | ENST00000456279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008165.hg.1 | NR_003950.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000412091.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000424928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000424928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000429320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000441664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000441664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013571.hg.1 | ENST00000450353.1 | Ensembl |

ZDHHC8P1
Genomic Map