Assay Details
| Gene Symbol: LOC105377962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134602.1 | 3 - 4 | 277 | 108 | ||
| GenBank mRNA | AI215976.1 | - | 1 - 2 | 108 | ||
| CR741031.1 | - | 3 - 4 | 279 | 108 | ||
More Information
Set Membership: |
|

LOC105377962
Genomic Map