Assay Details
| Gene Symbol: EPM2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011300.1 | XP_016866789.1 | 1 - 2 | 177 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011300.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |

EPM2A
Genomic Map