Assay Details
| Gene Symbol: MPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310934.1 | - | 1 - 2 | 141 | 100 | |
| DC402302.1 | - | 1 - 2 | 214 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310934.1 | - | 1 - 2 | 141 | 100 | |
| DC402302.1 | - | 1 - 2 | 214 | 100 | ||
Set Membership: |
|