| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000582052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000582052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000304046.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000394169.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000579695.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000581284.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000582052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000582052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
ENST00000584220.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010592.hg.1 |
NM_139280.3 |
Refseq |