Assay Details
| Gene Symbol: LOC100129935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026870.1 | 1 - 2 | 1287 | 90 | ||
| GenBank mRNA | AK023628.1 | - | 1 - 2 | 1288 | 90 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000594263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | ENST00000597791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008073.hg.1 | NR_026870.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011732.hg.1 | ENST00000599317.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008068.hg.1 | ENST00000601963.1 | Ensembl |

LOC100129935
Genomic Map