Assay Details
| Gene Symbol: FOSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114171.1 | NP_001107643.1 | 3 | 3543 | 85 | |
| NM_006732.2 | NP_006723.2 | 4 | 3651 | 85 | ||
| XM_005258691.1 | XP_005258748.1 | 5 | 3510 | 85 | ||
| GenBank mRNA | BC036724.1 | - | 5 | 3629 | 5212854 | 85 |
| BC040197.1 | - | 3 | 3651 | 4816584 | 85 | |
| D16366.1 | - | 1 | 117 | 85 | ||
| EU178109.1 | - | 4 | 3543 | 85 | ||
| EU178110.1 | - | 6 | 3510 | 85 | ||
| EU178111.1 | - | 4 | 3534 | 85 | ||
| EU178112.1 | - | 3 | 3426 | 85 | ||
| EU178113.1 | - | 5 | 3402 | 85 | ||
| EU178114.1 | - | 5 | 3393 | 85 | ||
| EU178115.1 | - | 3 | 3222 | 85 | ||
| EU178116.1 | - | 4 | 3081 | 85 | ||
| L49169.1 | - | 5 | 3651 | 85 | ||
More Information
Set Membership: |
|

FOSB
Genomic Map