| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
ENST00000368088.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
NM_004983.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
NM_004983.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
NM_004983.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
NM_004983.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010280.hg.1 |
NM_004983.2 |
Refseq |