| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000448199.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
ENST00000448199.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000418238.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000419536.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000429398.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000448199.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
ENST00000448199.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_037864.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009477.hg.1 |
NR_125965.1 |
Refseq |