Assay Details
| Gene Symbol: HHLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031693.2 | NP_001026863.1 | 2 - 3 | 398 | 75 | |
| NM_001036645.1 | NP_001031722.1 | 2 - 3 | 495 | 75 | ||
| NM_001036646.1 | NP_001031723.1 | 2 - 3 | 394 | 75 | ||
| XM_011540547.2 | XP_011538849.1 | 3 - 4 | 1648 | 75 | ||
| GenBank mRNA | AF126163.1 | - | 2 - 3 | 316 | 75 | |
| BC010922.1 | - | 2 - 3 | 376 | 4064190 | 75 | |
| BG773961.1 | - | 2 - 3 | 495 | 75 | ||
| BI868263.1 | - | 2 - 3 | 394 | 75 | ||
More Information
Set Membership: |
|

HHLA3
Genomic Map