Assay Details
| Gene Symbol: C1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123160.1 | - | 1 - 2 | 1494 | 168 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000343534.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000369718.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | ENST00000464591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300834.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_001300835.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009417.hg.1 | NM_174896.3 | Refseq |

C1orf162
Genomic Map