Assay Details
| Gene Symbol: CTTNBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL359565.1 | - | 1 - 2 | 560 | 133 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000607039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | ENST00000607039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000271277.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000441739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000502356.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000607039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | ENST00000607039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009441.hg.1 | NM_018704.2 | Refseq |

CTTNBP2NL
Genomic Map