Assay Details
Gene Symbol: WDR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC128385.1 | - | 2 - 3 | 116 | 62 | |
BC128454.1 | - | 2 - 3 | 116 | 62 | ||
BC148244.1 | - | 2 - 3 | 116 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
